Learn More
Sino Biological Anti-TIM-1/HAVCR1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80003RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat KIM-1 / TIM1 / HACVR1 (rR KIM-1 / TIM1 / HACVR1; Catalog#80003-R07H; NP_775172.1; Ser 18-Val 238). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1794