Learn More
Crystal Chem Inc Rat HDL-Cholesterol Assay Kit, 96-tests
SDP
Catalog No. 501947655
Encompass_Preferred
The HDL Cholesterol Rat Assay is used to quantify levels of HDL cholesterol in rat serum and plasma using only a 3 µL sample volume. Highlights: Kit uses sample size of only 3 µL. High sensitivity of 1.1 mg/dL. Dynamic range of 1.5 - 180 mg/dL. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7655 Supplier Crystal Chem Inc Supplier No. 79970
The HDL Cholesterol Rat Assay is used to quantify levels of HDL cholesterol in rat serum and plasma using only a 3 µL sample volume. Highlights: Kit uses sample size of only 3 µL. High sensitivity of 1.1 mg/dL. Dynamic range of 1.5 - 180 mg/dL. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.