Learn More
American Research Products Inc Rat heat shock protein 27 HSP
Catalog No. 5000326037
Encompass
Rat heat shock protein 27 HSP-27 ELISA Kit from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26037 Supplier American Research Products Inc Supplier No. CSBE09240R
Rat heat shock protein 27 HSP-27 ELISA Kit from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma tissue homogenates Method Sandwich