Learn More
American Research Products Inc Rat heat shock protein 70 hSP
Catalog No. 5000330128
Encompass
Rat heat shock protein 70 hSP-70 ELISA Kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-30128 Supplier American Research Products Inc Supplier No. CSBE08308R
Rat heat shock protein 70 hSP-70 ELISA Kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich