Learn More
American Research Products Inc Rat Heat shock protein HSP 90
Catalog No. 5000329440
Encompass
Rat Heat shock protein HSP 90-alpha (Hsp90aa1/Hsp86/Hspca) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29440 Supplier American Research Products Inc Supplier No. CSBE08309R
Rat Heat shock protein HSP 90-alpha (Hsp90aa1/Hsp86/Hspca) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich