Learn More
American Research Products Inc Rat heat shock protein HSP47
Catalog No. 5000326076
Encompass
Rat heat shock protein HSP47 ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-26076 Supplier American Research Products Inc Supplier No. CSBE11328R
Rat heat shock protein HSP47 ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich