Learn More
American Research Products Inc Rat heme oXygenase 1 HO 1 ELI
Catalog No. 5000329693
Encompass
Rat heme oXygenase 1 HO-1 ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29693 Supplier American Research Products Inc Supplier No. CSBE08267R
Rat heme oXygenase 1 HO-1 ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich