Learn More
Crystal Chem Inc Rat Hemoglobin A1c (HbA1c) Assay Kit, 96-tests
SDP
Catalog No. 501947686
Encompass_Preferred
The Rat Hemoglobin A1c Assay is used to quantify levels of rat HbA1c in whole blood using only a 5 µL sample volume.Highlights: Eliminates the need for total hemoglobin measurement. No interference from major hemoglobin variants including HbS, HbC, HbE. No interference from carbamylated Hb, acetylated Hb or Labile HbA1c. Excellent correlation to HPLC and Immunochemical methods. Designed for rat samples.Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7686 Supplier Crystal Chem Inc Supplier No. 80300
The Rat Hemoglobin A1c Assay is used to quantify levels of rat HbA1c in whole blood using only a 5 µL sample volume.Highlights: Eliminates the need for total hemoglobin measurement. No interference from major hemoglobin variants including HbS, HbC, HbE. No interference from carbamylated Hb, acetylated Hb or Labile HbA1c. Excellent correlation to HPLC and Immunochemical methods. Designed for rat samples.Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.