Learn More
American Research Products Inc Rat heparin binding EGF like g
Catalog No. 5000327831
Encompass
Rat heparin-binding EGF-like growth factor (HBEGF) ELISA kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27831 Supplier American Research Products Inc Supplier No. CSBEL010154RA
Rat heparin-binding EGF-like growth factor (HBEGF) ELISA kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma tissue homogenates Method Sandwich