Learn More
Raybiotech Inc Rat HGF ELISA Kit
Supplier:  Raybiotech Inc ELRHGF1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.8 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: rat 4-1BB, Activin A, Adiponectin, B7-1, CD48, CTACK, Decorin, Eotaxin, EphA5, Erythropoietin, FGF-BP, Fit-3 Ligand, Galectin-1, Galectin-3, Gas 1, GFR alpha-1, gp130, IL-1 R6, IL-1 ra, IL-2 R alpha, IL-3, IL-7, IL-17F, IL-22, JAM-A, MIP-1 alpha, Neuropilin-1, Neuropilin-2, Nope, Notch-1, Notch-2, P-Cadherin, Prolactin, RANTES, SCF, TIM-1, TIMP-2, TREM-1, TWEAK R.
Catalog No. 50-195-0250