Learn More
Sino Biological Anti-HGF Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80429T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat HGF / Hepatocyte Growth Factor (Catalog#80429-RNAH; NP_058713.1; Met1-Leu728). HGF / Hepatocyte Growth Factor specific IgG was purified by Rat HGF / Hepatocyte Growth Factor affinity chromatography.
Catalog No. 50-255-2772