Learn More
BIOHIPPO Rat HGFAC Microsample
SDP
Catalog No. 5000315829
Encompass
Rat HGFAC Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of HGFAC in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 6 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat HGFAC Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat HGFAC Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat HGFAC biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15829 Supplier BIOHIPPO Supplier No. BHE15210840
Rat HGFAC Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of HGFAC in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 6 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat HGFAC Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat HGFAC Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat HGFAC biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color