Learn More
American Research Products Inc Rat High density lipoprotein
Catalog No. 5000326144
Encompass
Rat High density lipoprotein HDL ELISA Kit from CUSABIO Detection Range 78 ng/mL-5000 ng/mL Sensitivity 19 5 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26144 Supplier American Research Products Inc Supplier No. CSBE11703R
Rat High density lipoprotein HDL ELISA Kit from CUSABIO Detection Range 78 ng/mL-5000 ng/mL Sensitivity 19 5 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich