Learn More
American Research Products Inc Rat High density lipoprotein c
Catalog No. 5000329460
Encompass
Rat High density lipoprotein cholesterol HDL-C ELISA kit from CUSABIO Detection Range 93 75 ng/mL-6000 ng/mL Sensitivity 23 4 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29460 Supplier American Research Products Inc Supplier No. CSBE14399R
Rat High density lipoprotein cholesterol HDL-C ELISA kit from CUSABIO Detection Range 93 75 ng/mL-6000 ng/mL Sensitivity 23 4 ng/mL Sample serum plasma tissue homogenates Method Sandwich