Learn More
Biomatik Corporation Rat Histone Cluster 1, H4a (HIST1H4A) ELISA Kit
Supplier:  Biomatik Corporation EKU10984
Encompass_Preferred
Rat Histone Cluster 1, H4a (HIST1H4A) ELISA Kit for detecting HIST1H4A in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.069ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P62804. Gene ID: 291152. Target Synonyms: H4FA; HIST2H4B; HIST2H4C; HIST2H4D; HIST2H4E; HIST2H4F; HIST2H4G; HIST2H4H; HIST2H4I; HIST2H4J; HIST2H4K; HIST2H4B; HIST4H4; HIST2H4A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6558