Learn More
Biomatik Corporation Rat Histone Deacetylase 1 (HDAC1) ELISA Kit
Supplier:  Biomatik Corporation EKU04813
Encompass_Preferred
Rat Histone Deacetylase 1 (HDAC1) ELISA Kit for detecting HDAC1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.28ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q4QQW4. Gene ID: 297893. Target Synonyms: HDAC1; GON10; HD1; RPD3; RPD3L1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5512