Learn More
Biomatik Corporation Rat Histone Deacetylase 2 (HDAC2) ELISA Kit
Supplier:  Biomatik Corporation EKU08478
Encompass_Preferred
Rat Histone Deacetylase 2 (HDAC2) ELISA Kit for detecting HDAC2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: F7ENH8. Gene ID: 84577. Target Synonyms: RPD3; YAF1; HD2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6753