Learn More
BIOHIPPO Rat hs-cTnT Microsample
SDP
Catalog No. 5000313724
Encompass
Rat hs-cTnT Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of hs-cTnT in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 5 3 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat hs-cTnT Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat hs-cTnT Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat hs-cTnT biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13724 Supplier BIOHIPPO Supplier No. BHE15212366
Rat hs-cTnT Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of hs-cTnT in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 5 3 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat hs-cTnT Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat hs-cTnT Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat hs-cTnT biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color