Learn More
American Research Products Inc Rat hypoXia inducible factor 1
Catalog No. 5000329623
Encompass
Rat hypoXia-inducible factor 1a HIF-1a ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29623 Supplier American Research Products Inc Supplier No. CSBE08540R
Rat hypoXia-inducible factor 1a HIF-1a ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich