Learn More
Biomatik Corporation Rat Hypoxia Inducible Factor 1 Alpha (HIF1a) CLIA Kit
Supplier:  Biomatik Corporation EKU09506
Encompass_Preferred
Rat Hypoxia Inducible Factor 1 Alpha (HIF1a) CLIA Kit for detecting HIF1a in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 27.4-20,000pg/ml. Sensitivity: 9.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O35800. Gene ID: 29560. Target Synonyms: HIF1-A; MOP1; PASD8; Basic Helix-Loop-Helix Transcription Factor; ARNT-interacting protein; Basic-helix-loop-helix-PAS protein MOP1; Class E basic helix-loop-helix protein 78. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7543