Learn More
Sino Biological Rat ICAM-1/CD54 Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000786010
Encompass_Preferred
The Rat ICAM-1 / CD54 ELISA Pair Set is for the quantitative determination of Rat ICAM-1 / CD54. This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for ICAM-1 / CD54 coated on a 96-well plate. Standards and samples are added to the wells, and any ICAM-1 / CD54 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-ICAM-1 / CD54 monoclonal antibody is then added, producing an antibody-antigen-antibody “sandwich”. The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of ICAM-1 / CD54 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86010 Supplier Sino Biological Supplier No. SEK8002215
The Rat ICAM-1 / CD54 ELISA Pair Set is for the quantitative determination of Rat ICAM-1 / CD54. This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for ICAM-1 / CD54 coated on a 96-well plate. Standards and samples are added to the wells, and any ICAM-1 / CD54 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-ICAM-1 / CD54 monoclonal antibody is then added, producing an antibody-antigen-antibody “sandwich”. The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of ICAM-1 / CD54 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.