Learn More
Abcam Rat ICAM1 ELISA Kit (CD54), 1 x 96 Tests
Supplier:  Abcam AB1007631X96TESTS
Encompass_Preferred
Rat ICAM1 ELISA Kit (CD54) is a Sandwich (quantitative) ELISA kit for the measurement of Rat ICAM1 (CD54) in Rat in Plasma, Cell culture supernatant, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2743349