Learn More
Sino Biological Recombinant Anti-ICAM-1/CD54 Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 80022R037A100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat ICAM-1 / CD54 (rR ICAM-1 / CD54; Catalog#80022-R08H; Q00238-1; Met1-Thr493) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-240-8592