Learn More
Sino Biological Anti-ICAM-1/CD54 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 80022MM15100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat ICAM-1 (rR ICAM-1; Catalog#80022-R08H; Q00238-1; Met1-Thr493). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-9349