Learn More
Sino Biological Anti-ICAM-1/CD54 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80022T60100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat ICAM-1/CD54 (rR ICAM-1/CD54; Catalog#80022-R08H; Q00238-1; Met1-Thr493). ICAM-1/CD54 specific IgG was purified by affinity chromatography.
Catalog No. 50-241-1018