Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat IFN-alpha 2 Recombinant Protein

Catalog No. PI131001
Change view
Click to view available options
Quantity:
1 x 10^5 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI131001 1 x 10^5 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI131001 Supplier Invitrogen™ Supplier No. 131001
Only null left
Add to Cart
Add to Cart

Recombinant Protein

Each blood donation used for interferon production tested negative for: HBsAg, HIV I-II, HCV and VDRL. After receipt, this product should be kept at 4°C while lyophilized and at -70°C after reconstitution in sterile distilled water for retention of full activity. After reconstitution store at -70°C in aliquots. When thawing, the contents of the tube should be apportioned in separate tubes so that freezing and thawing is kept to a minimum. Further dilute only in buffers containing protein such as 0.1% bovine serum albumin (BSA).

IFN alpha (Interferon alpha) is a Type I interferon belonging to a family of structurally related, species-specific proteins exclusive to vertebrates. The interaction of IFN-alpha with its receptor component result in the activation of a number of signaling pathways which are mainly involved in innate immune response against viral infection. In humans, the gene for IFN alpha is located on on chromosome 9. The type I interferons consist of 14 different alpha isoforms (subtypes with slightly different specificities), and single beta, omega, epsilon and kappa isoforms. Homologous molecules are found in many species, including rats and mice (and most mammals), and have been identified in birds, reptiles and fish species. IFN alpha is produced primarily by plasmacytoid dendritic cells, is a potent component of the anti-viral innate immune response and modulates adaptive immunity. Diseases associated with the dysfunction of IFN alpha include Hepatitis C and Hepatitis.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) ELISA, Immunoprecipitation, Western Blot
Formulation citric acid with 0.125 M trehalose, NaCl and no preservative; pH 5
Gene ID (Entrez) 298213
Molecular Weight (g/mol) 19 kDa
Name Rat IFN-alpha 2
pH Range 5
Quantity 1 x 10^5 units
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias alpha-2 A interferon; Ifa2; IFNA; IFN-a; IFNA 2; Ifna2; IFNA2A; IFNA2B; IFNA2C; IFN-alpha 1 b; IFN-alpha-1/13; IFN-alpha-2; IFN-alphaA; INFA2; Interferon; interferon alpha 1 b; interferon alpha 2; interferon alpha 2 A; interferon alpha 2 b; interferon alpha A; interferon alpha family, gene 2; interferon alpha-2; Interferon alpha-A; interferon alpha-D; interferon, alpha 2; Interferon-alpha II; leIF A; leIF D; RP11-354P17.2
Common Name Interferon alpha-2
Gene Symbol Ifna2
Biological Activity EC50 ̃ 1 U/mL; determined by use of the cytopathic effect inhibition assay described in [Rubinstein, S. et al. (1981). J. Virol. 37, 755-758 ]; [Familletti, P.C. et al. (1981). Methods in Enzymology, Vol. 78, 387-394 ]As there is no international Rat IFN-alpha standard, units are determined with respect to the international reference standard for mouse interferon alpha/beta (mouse IFN-alpha/beta) provided by the National Institutes of Health.
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Content And Storage 4°C
Expression System E. coli
Form Lyophilized
Protein Subtype Recombinant
Purity or Quality Grade >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.