Learn More
FUJIFILM BIOSCIENCES INC Rat IFN-gamma, Recombinant Protein, Shenandoah Biotechnology, 100 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 30004100UG
Encompass_Preferred
Interferon gamma (IFN-Y) is a type II interferon that is critical during adaptive and innate immune responses to infection.  IFN-Y is produced by T cells and natural killer cells following antigen-specific activation.  IFN-Y binds IFN-Y receptors (IFN-Y R1 and IFN-Y R2), which are expressed on most immune cells, to activate the JAK-STAT pathway.  IFN-Y-induced signaling increases the expression of class 1 major histocompatibility complex (MHC) molecules.  
Catalog No. 50-990-905
Edge