Learn More
Sino Biological Recombinant Anti-interferon-alpha/IFNA5 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 80205R003100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat IFNA5 / IFNaG (rR IFNA5 / IFNaG; Catalog#80205-R08H; XP_001076062.1; Met1-Glu189).
Catalog No. 50-241-2203