Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat IgE ELISA Kit

Catalog No. EEL126
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEL126 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEL126 Supplier Invitrogen™ Supplier No. EEL126
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Rat IgE ELISA quantitates IgE in serum, plasma, and other biological fluids The Rat IgE solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the binding of the second (detector) antibody to the target on a different epitope from the capture antibody. An antibody conjugated with enzyme binds the formed sandwich. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Rat IgE, also referred to as rat immunoglobulin E, is an antibody isotype synthesized by rats in response to allergens or parasites. It plays a pivotal role in mediating allergic reactions and immune responses against specific pathogens. IgE antibodies are primarily responsible for eliciting the release of inflammatory mediators, such as histamine, from mast cells and basophils. This immune response can give rise to symptoms like pruritus, edema, and bronchoconstriction. Investigating the genetic target Rat IgE can provide valuable insights into the underlying mechanisms of allergic diseases and contribute to the development of innovative therapeutic strategies for allergies and immunological disorders in both rats and humans.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01855
Assay Range 1.56-100 ng/mL
Assay Sensitivity 0.94 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Immunoglobulin
Sample Type Plasma, Serum
Target Species Rat
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Immunoglobulin E; ImmunoglobulinEImmunoglobulin E; ImmunoglobulinE;
Gene Symbol IGHE
Interassay CV <10%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 100 μL, Serum 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Refer to product documentation for component specific storage temperature.
Target Rat IgE
Test Time 1 hr 20 min
Total Assay Time 3 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.