Learn More
Crystal Chem Inc Rat IGFBP-2 ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80578
Encompass
The Rat IGFBP-2 ELISA is used to quantify levels of rat IGFBP-2 in serum using only a 5 µL sample volume. Highlights: Kits use only 5 µL sample, ideal for rat applications. Dynamic range of 31.25 - 2000 pg/mL. High sensitivity of 0.01 ng/mL.Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7745