Learn More
Raybiotech Inc Rat IGFBP-5 ELISA Kit
Supplier:  Raybiotech Inc ELRIGFBP51
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 25 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects rat IGFBP-5 and mouse IGFBP-5.
Catalog No. 50-195-0254