Learn More
Chondrex Inc RAT IGG α COL2 ELISA, 1 KIT
Catalog No. 501526705
Encompass_Preferred
ELISA kit designed to assay rat IgG antibodies against rat type II collagen in rat serum and plasma samples. This kit includes a 96-well ELISA plate with removable strips (10 bovine type II collagen coated strips and 4 standard coated strips), as well as all reagents necessary to complete the assay. Chromagen: TMB. Detection Range: 16 units/mL - 0.25 units/mL (82 units is approximately 1 µg IgG Ab/ml). Working Time: 4.25 hours. Ideal for quantifying immune response to autologous rat type II collagen after induction of Collagen Induced Arthritis (CIA) model in rats.
Catalog No. 50-152-6705 Supplier Chondrex Inc Supplier No. 2044T
ELISA kit designed to assay rat IgG antibodies against rat type II collagen in rat serum and plasma samples. This kit includes a 96-well ELISA plate with removable strips (10 bovine type II collagen coated strips and 4 standard coated strips), as well as all reagents necessary to complete the assay. Chromagen: TMB. Detection Range: 16 units/mL - 0.25 units/mL (82 units is approximately 1 µg IgG Ab/ml). Working Time: 4.25 hours. Ideal for quantifying immune response to autologous rat type II collagen after induction of Collagen Induced Arthritis (CIA) model in rats.