Learn More
Innovative Research Inc Rat IgG Depleted Srm 100ml
Catalog No. 502744006
Encompass_Preferred
Rat IgG Depleted Serum 100ml - Rat IgG Depleted Serum from Innovative Research has been purified from normal rat serum by affinity chromatography. This is a Frozen liquid. This product can be used as a control in applications like ELISA and Western Blot. It can be used for a wide range of in vitro applications, including tissue culture and more.
Catalog No. 50-274-4006 Supplier Innovative Research Inc Supplier No. IRTIGGDS100ML
Rat IgG Depleted Serum 100ml - Rat IgG Depleted Serum from Innovative Research has been purified from normal rat serum by affinity chromatography. This is a Frozen liquid. This product can be used as a control in applications like ELISA and Western Blot. It can be used for a wide range of in vitro applications, including tissue culture and more.