Learn More
Rockland Immunochemicals Rat IgG (H&L) Antibody ATTO 647N Conjugated Pre-Adsorbed 500 µg
SDP
Supplier:  Rockland Immunochemicals 612156120
Encompass
Anti-Rat IgG (H&L) conjugated to ATTO 647N is designed for STED microscopy FRET immunofluorescence microscopy fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis including multicolor imaging utilizing various commercial platforms.
Catalog No. 50-194-3933