Learn More
Abcam Rat IgG ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1895781X96TESTS
Encompass_Preferred
Rat IgG ELISA Kit is a single-wash 90-min Simplestep used to quantify Rat IgG with a sensitivity of 55 pg/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10x96 plates for bulk orders and 384-well plate for higher throughput - Cited in over 10 citations
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0926098