Learn More
AdipoGen Rat IgG2a Isotype Ctrl A488
SDP
Supplier:  AdipoGen AG35B0002TDC050
Encompass_Preferred
Monoclonal Antibody. Isotype Rat IgG2akappa. Clone KLH/G2a-11. Applications FACS, ICC, IHC, IP. Liquid. In PBS containing 0.02% sodium azide. Negative control in assays where rat IgG2a ATTO 488-labeled antibodies are used.
Catalog No. 50-256-3095