Learn More
AdipoGen Rat IgG2b Isotype Ctrl
SDP
Supplier:  AdipoGen AG35B0011C050
Encompass_Preferred
Monoclonal Antibody. Isotype Rat IgG2bkappa. Clone KLH/G2b-1-2. Applications FACS, ICC, IHC, IP. Liquid. In PBS containing 0.02% sodium azide. Negative control in assays where rat IgG2b antibodies are used.
Catalog No. 50-256-3080