Learn More
STEMCELL Technologies Rat IgM, kappa Isotype Control Antibody, Clone RTK2118, PE, Size: 100 µg
SDP
Supplier:  STEMCELL Technologies 60074PE
Encompass
Clone RTK2118 is a rat monoclonal antibody suitable for use as an isotype-matched control antibody in several applications to estimate the degree of non-specific binding by an antigen-specific antibody. This antibody clone has been verified for use as an isotype control antibody for assessing non-specific binding to cells in applications such as flow cytometry and immunofluorescence microscopy applications (surface and intracellular staining). The use of an appropriate isotype control helps confirm the specificity of the antigen-specific antibody and indicates non-specific binding that may result from binding to Fc receptors or other cell components. Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5425
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov