Learn More
Raybiotech Inc Rat IL-10 ELISA Kit
Supplier:  Raybiotech Inc ELRIL10CL1
Encompass_Preferred
For cell and tissue lysate samples. Sensitivity: 10 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes rat IL-10.
Catalog No. 50-195-0256