Learn More
Bon Opus Biosciences Rat Interleukin-12/IL-12(C-His) -50ug
SDP
Supplier:  Bon Opus Biosciences BP15350UG
Encompass_Preferred
Recombinant Rat Interleukin-12 is produced by our Mammalian expression system and the target gene encoding Met23-Ser335(p40)/Arg23-Ser215(p35) is expressed with a 6His tag at the C-terminus. Expression system: Human 293 Cells. Accession# Q9R278/Q9R103. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# BP153
Catalog No. 50-208-0241