Learn More
Gibco™ Rat IL-13 (113 aa) Recombinant Protein, PeproTech®
Description
Recombinant Rat IL-13 a 12.3 kDa protein consisting of 113 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | P42203 |
| For Use With (Application) | Control, Western Blot Control, Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 116553 |
| Molecular Weight (g/mol) | 12.3 kDa |
| Name | Rat IL-13 (113 aa) |
| Purification Method | Purified |
| Quantity | 100 μg |
| Source | E. coli |
| Storage Requirements | -20°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.