Learn More
Raybiotech Inc Rat IL-18 ELISA Kit
Supplier:  Raybiotech Inc ELRIL181
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 1.6 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Rat IL-18
Catalog No. 50-195-0259