Learn More
ACROBiosystems ACROBiosystems Rat IL-2 R beta&IL-2 R alpha&IL-2 R gamma Protein, His Tag&Twin-Strep Tag
Catalog No. 502106440
Encompass_Preferred
Recombinant Protein;1MG;Rat IL-2RB&IL-2RA&IL-2RG, His Tag&Twin-Strep Tag (ILG-R5223) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Glu 239 (IL-2RB) & Glu 22 - Gln 235 (IL-2RA) & Ser 25 - Ala 262 (IL-2RG) (Accession # P26896-1 (IL-2RB) & P26897-1 (IL-2RA) & Q68FU6-1 (IL-2RG)).;Rat IL-2RB&IL-2RA&IL-2RG, His Tag&Twin-Strep Tag is produced by co-expression of IL-2RB and IL-2RA and IL-2RG, has a calculated MW of 29.3 kDa (IL-2RB) and 58.2 kDa (IL-2RA&IL-2RG). Subunit IL-2RB is fused with a his tag at the C-terminus and subunit IL-2RA&IL-2RG is fused with a Twin-Strep Tag at the C-terminus.
Catalog No. 50-210-6440 Supplier ACROBiosystems Supplier No. ILGR52231MG
Recombinant Protein;1MG;Rat IL-2RB&IL-2RA&IL-2RG, His Tag&Twin-Strep Tag (ILG-R5223) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Glu 239 (IL-2RB) & Glu 22 - Gln 235 (IL-2RA) & Ser 25 - Ala 262 (IL-2RG) (Accession # P26896-1 (IL-2RB) & P26897-1 (IL-2RA) & Q68FU6-1 (IL-2RG)).;Rat IL-2RB&IL-2RA&IL-2RG, His Tag&Twin-Strep Tag is produced by co-expression of IL-2RB and IL-2RA and IL-2RG, has a calculated MW of 29.3 kDa (IL-2RB) and 58.2 kDa (IL-2RA&IL-2RG). Subunit IL-2RB is fused with a his tag at the C-terminus and subunit IL-2RA&IL-2RG is fused with a Twin-Strep Tag at the C-terminus.