Learn More
Raybiotech Inc RAT IL-6 ELISA
Supplier:  Raybiotech Inc ELRIL61
Encompass_Preferred
RayBio Rat IL-6 ELISA Kit for cell culture supernatants, plasma, and serum samples. Pre-coated 96-well microplate kit with additional reagents. Detection Range: 30 pg/ml - 10000 pg/ml
Catalog No. 50-104-4951