Learn More
Raybiotech Inc Rat IL-7 ELISA Kit
Supplier:  Raybiotech Inc ELRIL71
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 8 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Rat IL-7
Catalog No. 50-195-0271