Learn More
Invitrogen™ Rat IL-9 ELISA Kit
Description
Rat IL-9 quantitates rat IL-9 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat IL-9.
Specifications
Specifications
| Accession Number | cytokine P40; homolog of mouse T cell and mast cell growth factor 40; HP40; Il9; IL-9; ILN; Interleukin; interleukin 9; Interleukin9; interleukin-9; P40; p40 cytokine; p40 T-cell and mast cell growth factor; T-cell growth factor p40 |
| Assay Range | 0.45-100 ng/mL |
| Assay Sensitivity | 0.45 ng/mL |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Rat |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Gene Alias | cytokine P40; homolog of mouse T cell and mast cell growth factor 40; HP40; IL9; IL-9; ILN; Interleukin; interleukin 9; Interleukin9; interleukin-9; P40; p40 cytokine; p40 T-cell and mast cell growth factor; T-cell growth factor P40cytokine P40; homolog of mouse T cell and mast cell growth factor 40; HP40; IL9; IL-9; ILN; Interleukin; interleukin 9; Interleukin9; interleukin-9; P40; p40 cytokine; p40 T-cell and mast cell growth factor; T-cell growth factor P40; |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.