Learn More
Sino Biological Anti-IL-10 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80082T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat IL-10 (rR IL-10; Catalog#80082-RNAE; NP_036986.2; Ser19-Asn178). IL-10 specific IgG was purified by Rat IL-10 affinity chromatography.
Catalog No. 50-241-1935