Learn More
BIOHIPPO Rat IL11Ra Microsample
SDP
Catalog No. 5000315061
Encompass
Rat IL11Ra Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL11Ra in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 063 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat IL11Ra Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat IL11Ra Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat IL11Ra biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15061 Supplier BIOHIPPO Supplier No. BHE15210944
Rat IL11Ra Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL11Ra in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 063 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat IL11Ra Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat IL11Ra Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat IL11Ra biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color