Learn More
Sino Biological Anti-IL-12 p40/IL12B Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80352T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat IL12B / IL-12B (Catalog#80352-R08H; G3V9Y5; Met1-Ser335). IL12B / IL-12B specific IgG was purified by Rat IL12B / IL-12B affinity chromatography.
Catalog No. 50-241-1333