Learn More
Sino Biological Rat IL12B Protein 50ug (Sino Biological)
Supplier:  Sino Biological 10357MM0220
Encompass_Preferred
A DNA sequence encoding the rat Il12b (XP_006246209.1) (Met1-Ser335) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-4388